- Effects of chloride, bromide, and iodide upon decomposition of nucleosides induced by ultrasound in neutral solution.
Effects of chloride, bromide, and iodide upon decomposition of nucleosides induced by ultrasound in neutral solution.
Nucleosides, nucleotides & nucleic acids (2010-07-28)
Toshinori Suzuki, Kenji Yamada, Michiyo Inukai
PMID20661814
ABSTRAKT
When the ultrasound of 42 kHz was irradiated on a neutral mixture of 2'-deoxycytidine, 2'-deoxyguanosine, 2'-deoxythymidine, and 2'-deoxyadenosine, concentrations of all the nucleosides decreased. Addition of NaCl to the system had no effect. NaBr suppressed the reactions for all the nucleosides, but the efficiency of 2'-deoxyguanosine was low. NaI suppressed the reactions for all the nucleosides more effectively. A comparison with the results of the effects of halides on the reaction of nucleosides by a Fenton system suggested that only half of the nucleoside damage in the ultrasound-irradiated solution was caused by hydroxyl radicals formed from water by the sonication.