Merck
Quenching of triplet benzophenone by benzene and diphenyl ether: a DFT study.
The journal of physical chemistry. A (2010-09-16)
Margaret J Smith, Götz Bucher
ABSTRAKT
The reaction of triplet benzophenone with benzene and diphenyl ether has been studied by density functional theory. Quenching of the triplet ketone is predicted to occur by addition of the carbonyl oxygen to the arene chromophores. The reaction is accompanied by a significant degree of charge transfer. In case of the reaction of triplet benzophenone with diphenyl ether (DPE), addition is predicted to occur preferentially at the ortho position of the DPE molecule. Addition to the ipso-position of DPE, which provides a pathway for formation of the phenoxy radical, is predicted to occur as a minor reaction pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diphenyl ether, ≥99%, FG
Supelco
Diphenyl ether, Selectophore, ≥99.9%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, ≥99%