Merck
An in vitro comparison of a new vinyl chalcogenide and sodium selenate on adenosine deaminase activity of human leukocytes.
Chemico-biological interactions (2010-12-22)
Luziane Potrich Bellé, Paula Eliete Rodrigues Bitencourt, Faida Husein Abdalla, Robson Brum Guerra, Claudia Funchal, Maria Beatriz Moretto
ABSTRAKT
Selenium (Se) is a dietary essential trace element with important biological roles. Sodium selenate (Na(2)SeO(4)) is an inorganic Se compound used in human and animal nutrition that acts as precursor for selenoprotein synthesis. The organoselenium 3-methyl-1-phenyl-2-(phenylseleno)oct-2-en-1-one (C(21)H(2)HOSe) is an α,β-unsaturated ketone functionalized vinyl chalcogenide that has been found as a potential tool in organic synthesis. Adenosine deaminase (ADA) is an important enzyme in the degradation of adenine nucleotides. In this study, we investigated the in vitro effects of both Se compounds on ADA activity and cell viability in leukocyte suspension (LS) of healthy donors (n=12). We first observed an inhibition of ADA activity using 0.1 μM of 3-methyl-1-phenyl-2-(phenylseleno)oct-2-en-1-one, and an increase in cellular viability when 30 μM were used. However, we did not observe alterations in the presence of sodium selenate. Moreover, both Se compounds did not alter lactate dehydrogenase activity and thiobarbituric acid reactive substance levels. These results suggest that the inhibition of ADA activity caused by α,β-unsaturated ketone may affect the adenosine levels in LS and modulate cell viability, attenuating conditions that involve the activation of the immune system.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium selenate, BioXtra
Sigma-Aldrich
Sodium selenate, ≥95% (elemental analysis)