Merck
  • [Simultaneous determination of delta-9-tetrahydrocannabinol cannabidiol and cannabinol in edible oil using ultra performance liquid chromatography-tandem mass spectrometry].
[Simultaneous determination of delta-9-tetrahydrocannabinol cannabidiol and cannabinol in edible oil using ultra performance liquid chromatography-tandem mass spectrometry].
Se pu = Chinese journal of chromatography (2011-03-09)
Aizhi Zhang, Quanlin Wang, Shijie Mo
ABSTRAKT
A method for the simultaneous determination of delta-9-tetrahydrocannabinol (THC), cannabidiol (CBD) and cannabinol (CBN) in edible oil was developed using ultra performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS). The target compounds were extracted with methanol, purified by an LC-Alumina-N solid phase extraction cartridge, separated and detected by the UPLC-MS/MS. Quantitative analysis was corrected by an isotope internal standard method using delta-9-THC-D3 as internal standard. Average recoveries for the target compounds varied from 68.0% to 101.6% with the relative standard deviations ranging from 7.0% to 20.1% at three spiked levels. The limits of detection (LOD) of the method were from 0.06-0.17 microg/kg and the limits of quantification (LOQ) were in the range of 0.20-0.52 microg/kg. The results showed that the method is able to meet the requirements for the simultaneous determination of THC, CBD and CBN in edible oil.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cannabinol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Δ9-Tetrahydrocannabinolic acid A, analytical standard