Merck
The antiangiogenic and antinociceptive activities of n-propyl gallate.
Phytotherapy research : PTR (2011-03-29)
H-J Jung, C-J Lim
ABSTRAKT
n-Propyl gallate dose-dependently displayed an inhibitory effect on chick chorioallantoic membrane (CAM) angiogenesis. It markedly increased the endostatin level in both isolated CAM tissues and human umbilical vein endothelial cells (HUVECs). n-Propyl gallate was also able to enhance the endostatin mRNA level in HUVECs. Antinociceptive activity of n-propyl gallate was assessed using an acetic acid-induced writhing test in mice. In brief, n-propyl gallate possesses antiangiogenic activity via up-regulation of endostatin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98.0% (HPLC)
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propyl gallate, ≥98%, FCC