Merck
  • Expression profiles of gonadotropins and their receptors during 17α-methyltestosterone implantation-induced sex change in the orange-spotted grouper (Epinephelus coioides).
Expression profiles of gonadotropins and their receptors during 17α-methyltestosterone implantation-induced sex change in the orange-spotted grouper (Epinephelus coioides).
Molecular reproduction and development (2011-05-14)
Xuesong Hu, Xiaochun Liu, Haifa Zhang, Yong Zhang, Shuisheng Li, Qing Sang, Qian Wang, Wenna Luo, Qizhi Liu, Danqi Lu, Zining Meng, Haoran Lin
ABSTRAKT
It is known that the hypothalamic-pituitary-gonadal axis participates in the sex change of hermaphrodite teleosts, and gonadal steroid hormones mediate this physiological process. The secretion of gonadal steroids is directly regulated by signaling pathways involving gonadotropins (GtHs) and gonadotropin receptors (GtHRs) in teleosts. To gain insight into the involvement of GtH/GtHR systems in the sex change process, cDNAs encoding follicle-stimulating hormone receptor (FSHR) and luteinizing hormone receptor (LHR) were firstly isolated from gonads of orange-spotted grouper (Epinephelus coioides), a protogynous hermaphrodite fish. Reverse transcription-PCR (RT-PCR) analysis demonstrated that the expression of the FSHR was confined to the brain, pituitary gland, ovary, and testis, while the LHR was expressed only in the brain, ovary, and testis. Furthermore, the expression profiles of GtH subunits (FSHβ and LHβ) and their receptors were analyzed in parallel with the serum levels of estradiol-17β (E(2) ), testosterone (T), and 11-ketotestosterone (11-KT) during 17α-methyltestosterone (MT)-induced sex change. Quantitative real-time PCR determined that the abundances of FSHβ and FSHR were significantly inhibited after MT treatment for 2 and 4 weeks, but subsequently returned to the control level after 6 weeks. In contrast, the mRNA levels of LHβ and LHR were significantly elevated throughout the sex change process. During MT-induced sex change, serum concentrations of E(2) remained constant while T and 11-KT levels were significantly increased. Taken together, our results suggest that GtH/GtHR systems are involved in MT-induced sex change, and two signaling pathways may have distinct roles in modulating the variations of the corresponding steroid hormones in the orange-spotted grouper.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
17α-Methyltestosterone, ≥97.0% (HPLC)
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Supelco
17α-Methyltestosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®