Merck
  • Multiple pathways from three types of sugar receptor sites to metabotropic transduction pathways of the blowfly: study by the whole cell-clamp experiments.
Multiple pathways from three types of sugar receptor sites to metabotropic transduction pathways of the blowfly: study by the whole cell-clamp experiments.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2011-06-01)
Hideko Kan, Naoko Kataoka-Shirasugi, Taisaku Amakawa
ABSTRAKT
Multiple pathways from three types of multiple receptor sites to three types of metabotropic signal transduction pathways were investigated in the whole cell-clamp experiments using isolated labellar sugar receptor neurons (cells) of the adult blowfly, Phormia regina. First, the concentration-response curves of three types of sweet taste components specialized to multiple receptor sites were obtained: sucrose for the pyranose sites (P-sites), fructose for the furanose sites (F-sites), and l-valine for the alkyl sites (R-sites). Next, the effects of inhibitors such as 2', 5'-dideoxyadenosine on adenylyl cyclase in the cAMP pathway, LY 83583 on guanylyl cyclase in the cGMP pathway, and U-73122 on phospholipase C in the IP₃ pathway were examined. The results showed that all of the inhibitors affected each specific target in the second-messenger transduction pathways. The obtained results verified that the P-site corresponded to the cAMP, the F-site to the cGMP, and the R-site to the IP₃ transduction pathway, and that these three signal pathways did not have crossing points.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Anilinoquinoline-5,8-quinone, ≥95% (TLC), solid
Sigma-Aldrich
2′,5′-Dideoxyadenosine, ≥95% (HPLC), solid
Sigma-Aldrich
2′,3′-Dideoxyadenosine, ≥97% (HPLC)