Merck
Copper-catalyzed N-arylation of amines with part-per-million catalyst loadings under air at room temperature.
Chemical communications (Cambridge, England) (2011-07-06)
Ruilong Xie, Hua Fu, Yun Ling
ABSTRAKT
An efficient copper-catalyzed method for N-arylation of amines has been developed with part-per-million catalyst loadings at room temperature under air. Reactions of substituted (E)-1-(2-halophenyl)alkanone oximes with aliphatic amines or aromatic amines provided the N-arylation products in good to excellent yields.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium carbonate, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, ACS reagent, ≥99.0%