Merck
Hydrophilic flower-like CuS superstructures as an efficient 980 nm laser-driven photothermal agent for ablation of cancer cells.
Advanced materials (Deerfield Beach, Fla.) (2011-07-08)
Qiwei Tian, Minghua Tang, Yangang Sun, Rujia Zou, Zhigang Chen, Meifang Zhu, Shiping Yang, Jinglong Wang, Jianhua Wang, Junqing Hu
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Copper(II) sulfide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) sulfide, powder, −100 mesh, ≥99% trace metals basis