- Practical tip: Chicago Sky Blue (CSB) stain can be added to the routine potassium hydroxide (KOH) wet-mount to provide a color contrast and facilitate the diagnosis of dermatomycoses.
Practical tip: Chicago Sky Blue (CSB) stain can be added to the routine potassium hydroxide (KOH) wet-mount to provide a color contrast and facilitate the diagnosis of dermatomycoses.
Dermatology online journal (2011-09-13)
Christopher Seng-Hong Lim, Siew-Lin Lim
PMID21906491
ABSTRAKT
Rapid confirmation of dermatomycoses is desirable because it allows the clinician to initiate appropriate therapy without delay. The routine potassium hydroxide (KOH) wet-mount is cheap and rapid to use but this method lacks a color contrast. We offer a simple practical tip of adding Chicago Sky Blue (CSB) stain to KOH to highlight fungal elements and provide a color contrast that makes reading and interpretation simple, even for the novice.
MATERIAŁY