Merck
High-reactivity matrices increase the sensitivity of matrix enhanced secondary ion mass spectrometry.
Analytical chemistry (2011-09-24)
Fabian N Svara, Andras Kiss, Thorsten W Jaskolla, Michael Karas, Ron M A Heeren
ABSTRAKT
Secondary ion mass spectrometry (SIMS) is a desorption/ionization method in which ions are generated by the impact of a primary ion beam on a sample. Classic matrix assisted laser desorption and ionization (MALDI) matrices can be used to increase secondary ion yields and decrease fragmentation in a SIMS experiment, which is referred to as matrix enhanced SIMS (ME-SIMS). Contrary to MALDI, the choice of matrices for ME-SIMS is not constrained by their photon absorption characteristics. This implies that matrix compounds that exhibit an insufficient photon absorption coefficient have the potential of working well with ME-SIMS. Here, we evaluate a set of novel derivatives of the classical MALDI matrices α-cyano-4-hydroxycinnamic acid (CHCA) and 2,5-dihydroxybenzoic acid (DHB) for usability in ME-SIMS. This evaluation was carried out using peptide mixtures of different complexity and demonstrates significant improvements in signal intensity for several compounds with insufficient UV absorption at the standard MALDI laser wavelengths. Our study confirms that the gas-phase proton affinity of a matrix compound is a key physicochemical characteristic that determines its performance in a ME-SIMS experiment. As a result, these novel matrices improve the performance of matrix enhanced secondary ion mass spectrometry experiments on complex peptide mixtures.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gentisic acid sodium salt hydrate, ≥98%
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%