Merck
The 1,4-naphthoquinone derivative from Pyrola rotundifolia activates AMPK phosphorylation in C2C12 myotubes.
Fitoterapia (2011-10-01)
Leonid R Ptitsyn, Kenzo Nomura, Ilya V Sklyar, Anna B Ravcheeva
ABSTRAKT
An aqueous ethanol extract of Pyrola rotundifolia L. induced AMP-activated protein kinase (AMPK) phosphorylation in C2C12 myotubes. The bioassay-guided fractionation of the extract led to the isolation a 2-methyl-7-hydroxymethyl-1,4-naphthoquinone, or a 7'-hydroxy-chimaphilin, which showed concentration-dependent AMPK phosphorylation activity at 2.5-20 μg/ml. At a concentration of 10 μg/ml (50 μM), an approximately four-fold increase in the AMPKα(Thr¹⁷²) phosphorylation level was observed. The stimulatory effect of naphthoquinone on AMPK activity was comparable to that of known compounds found in natural sources that activate the AMPK signaling pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,4-Naphthoquinone, 97%
Sigma-Aldrich
1,4-Naphthoquinone, purum, ≥96.5% (HPLC)