Merck
Effects of olmesartan and imidapril on the plasma adiponectin, P-selectin, and MDA-LDL levels of diabetic nephropathy patients.
Clinica chimica acta; international journal of clinical chemistry (2011-10-11)
Kouichi Utsumi, Fumihiko Yasuda, Yoko Watanabe, Seiichiro Higo, Akio Hirama, Emiko Fujita, Kae Ueda, Akiko Mii, Tomohiro Kaneko, Masahiro Mishina, Yasuhiko Iino, Yasuo Katayama
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Imidapril hydrochloride, ≥98% (HPLC)