Merck
Metabolism studies of a small-molecule tumor necrosis factor-alpha (TNF-α) inhibitor, UTL-5b (GBL-5b).
European journal of drug metabolism and pharmacokinetics (2011-11-05)
Jiajiu Shaw, Brian Shay, Jack Jiang, Frederick Valeriote, Ben Chen
ABSTRAKT
UTL-5b is an anti-inflammatory and anti-arthritic small-molecule tumor necrosis factor-alpha inhibitor and a structural analogue of the anti-arthritic drug, leflunomide. Leflunomide is known to be metabolized to teriflunomide, but the metabolites of UTL-5b have not been reported. The objective of this study was to investigate whether UTL-5b has a similar metabolic behavior as leflunomide. Preliminary studies showed that when exposed to microsomes in vitro with or without NADPH, UTL-5b disappeared within 30 min. To further investigate the microsomal metabolism, liquid chromatography-ultraviolet (LC-UV) and LC/tandem mass spectrometry (LC-MS/MS) were employed to, respectively, monitor the microsomal metabolites and identify the structure of the metabolites using LC-full scan MS and LC combined with multiple-ion monitoring MS. Fragmentation determination was analyzed by two types of scans: product ion scans and precursor ion scan. The in vitro microsomal treatment of UTL-5b resulted in two major metabolites: 5-methylisoxazole-3-carboxylic acid and 2-chloroaniline. Thus, the in vitro metabolic behavior of UTL-5b appears to be different from that of leflunomide in that the isoxazole ring is cleaved.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Chloroaniline, ≥99.5% (GC)
Sigma-Aldrich
2-Chloroaniline, technical, ≥98.0% (GC)