Merck
Irritant-induced asthma.
Current opinion in allergy and clinical immunology (2012-02-14)
Manon Labrecque
ABSTRAKT
To describe the recent insights into the definition, causes, natural outcome, and key elements of irritant-induced asthma (IIA) management. IIA is a subtype of occupational asthma without immunologic sensitization and includes the typical reactive airway dysfunction syndrome (RADS) and a more gradual form called not-so-sudden IIA, when onset of asthma follows repeated low-dose exposure to irritants. The World Trade Center tragedy brought new insight in the understanding of IIA, suggesting that it can exhibit a prolonged interval between exposure and recognition of clinical symptoms and disease. Dimethyl sulfate has been recently reported to cause RADS and repeated diesel exhaust exposure to cause not-so-sudden IIA in patients who worked in a bus garage. Cleaning workers who are exposed to a large variety of irritants and sensitizers are especially at risk of occupational asthma and IIA. IIA includes RADS and not-so-sudden IIA. Outcome of IIA is as poor as occupational asthma with sensitization. Treatment of IIA does not differ from standard asthma treatment, but high-dose vitamin D could be assessed further for possible therapeutic benefit.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfate, ≥99.5%
Sigma-Aldrich
Dimethyl sulfate, ≥99%