Merck
A new orally active, aminothiol radioprotector-free of nausea and hypotension side effects at its highest radioprotective doses.
International journal of radiation oncology, biology, physics (2012-02-15)
Cheryl M Soref, Timothy A Hacker, William E Fahl
ABSTRAKT
A new aminothiol, PrC-210, was tested for orally conferred radioprotection (rats, mice; 9.0 Gy whole-body, which was otherwise lethal to 100% of the animals) and presence of the debilitating side effects (nausea/vomiting, hypotension/fainting) that restrict use of the current aminothiol, amifostine (Ethyol, WR-2721). PrC-210 in water was administered to rats and mice at times before irradiation, and percent-survival was recorded for 60 days. Subcutaneous (SC) amifostine (positive control) or SC PrC-210 was administered to ferrets (Mustela putorius furo) and retching/emesis responses were recorded. Intraperitoneal amifostine (positive control) or PrC-210 was administered to arterial cannulated rats to score drug-induced hypotension. Oral PrC-210 conferred 100% survival in rat and mouse models against an otherwise 100% lethal whole-body radiation dose (9.0 Gy). Oral PrC-210, administered by gavage 30-90 min before irradiation, conferred a broad window of radioprotection. The comparison of PrC-210 and amifostine side effects was striking because there was no retching or emesis in 10 ferrets treated with PrC-210 and no induced hypotension in arterial cannulated rats treated with PrC-210. The tested PrC-210 doses were the ferret and rat equivalent doses of the 0.5 maximum tolerated dose (MTD) PrC-210 dose in mice. The human equivalent of this mouse 0.5 MTD PrC-210 dose would likely be the highest PrC-210 dose used in humans. By comparison, the mouse 0.5 MTD amifostine dose, 400 μg/g body weight (equivalent to the human amifostine dose of 910 mg/m(2)), when tested at equivalent ferret and rat doses in the above models produced 100% retching/vomiting in ferrets and 100% incidence of significant, progressive hypotension in rats. The PrC-210 aminothiol, with no detectable nausea/vomiting or hypotension side effects in these preclinical models, is a logical candidate for human drug development to use in healthy humans in a wide variety of radioprotection settings, including medical radiation, space travel, and nuclear accidents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amifostine trihydrate, ≥97% (TLC), powder