Merck
  • Pyrvinium, a potent small molecule Wnt inhibitor, increases engraftment and inhibits lineage commitment of mesenchymal stem cells (MSCs).
Pyrvinium, a potent small molecule Wnt inhibitor, increases engraftment and inhibits lineage commitment of mesenchymal stem cells (MSCs).
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society (2012-02-16)
Sarika Saraswati, Desirae L Deskins, Ginger E Holt, Pampee P Young
ABSTRAKT
We and others have found that Wnt signaling inhibition is important in mesenchymal stem cell (MSC) self-renewal. Pyrvinium was identified as a potent Wnt inhibitor in a chemical screen for small molecules. In the present study, we hypothesized that pyrvinium will enhance MSC self-renewal to improve the clinical efficacy of MSC therapy. Pyrvinium increased MSC proliferation in vitro while inhibiting their osteogenic and chondrogenic lineage commitment by reducing cytoplasmic β-catenin. Although MSCs are a promising target for cell therapy, strategies to enhance their survival and maintain their stemness in the wounded area are essential. Using an in vivo model of granulation tissue formation, we demonstrated that pyrvinium enhanced long-term MSC engraftment. Pyrvinium-treated MSC-generated granulation tissue also demonstrated less ectopic differentiation into bone or cartilage. This study highlights the potential of using a therapeutic Wnt inhibitor to enhance MSC-driven regenerative therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)