Merck
  • Mechanistic study on the electrochemical reduction of 9,10-anthraquinone in the presence of hydrogen-bond and proton donating additives.
Mechanistic study on the electrochemical reduction of 9,10-anthraquinone in the presence of hydrogen-bond and proton donating additives.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2012-03-28)
Jiro Katsumi, Tatsushi Nakayama, Yukihiro Esaka, Bunji Uno
ABSTRAKT
The electrochemical reduction of 9,10-anthraquinone (AQ) was investigated in CH(3)CN in both the absence and presence of the hydrogen-bond and proton donating additives, CH(3)OH, CH(CF(3))(2)OH, phenol, 4-methoxyphenol, 4-cyanophenol, 2,4,6-trichlorophenol, and benzoic acid (BA). Three clearly different types of electrochemical behavior were observed with increasing concentrations of the additives, and were simulated to analyze the reaction mechanisms. Type I was observed for weakly interacting additives, such as CH(3)OH, characterized by positive shifts of the two well-separated reduction waves, corresponding to the formation of AQ(•-) and AQ(2-), with no loss of reversibility. The second wave shifted more strongly, and finally merged with the first. These behaviors are explained by the association of AQ(2-) with the additives via strong hydrogen-bonding. Type II is attributed to a reduction mechanism involving quantitative formation of strong hydrogen-bonded complexes of AQ(2-) with additives, such as CH(CF(3))(2)OH, phenol and 4-methoxyphenol, showing a reversible or quasireversible two-electron reduction wave with increasing concentrations of the additives. The behavior of Type III, observed in the presence of strongly interacting additives, such as 2,4,6-trichlorophenol and BA, is characterized by a voltammogram composed of the 2-electorn cathodic and the broad anodic waves without keeping reversibility, facilitated by proton transfer in the hydrogen-bonded complexes, AQ(•-)-BA and AQ(2-)-BA. The effects of hydrogen-bonding and protonation on the electrochemistry of AQ have been systematically demonstrated in terms of the potentials and reaction pathways of the various species, which appear in quinone-hydroquinone systems.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Melting point standard 283-286°C, analytical standard
Sigma-Aldrich
Anthraquinone, 97%