Merck
Enantioselective organocatalytic Michael addition of ketones to alkylidene malonates.
Chirality (2012-05-17)
Siang-En Syu, Chan-Hui Huang, Ko-Wei Chen, Chia-Jui Lee, Utpal Das, Yeong-Jiunn Jang, Wenwei Lin
ABSTRAKT
Organocatalysts bearing sulfide or sulfone functions (1a-d) were studied for the direct asymmetric Michael addition of ketones and alkylidene malonates. The organocatalyst (S)-2-((naphthalen-2-ylthio)methyl)pyrrolidine, bearing a pyrrolidine and a sulfide moiety, showed a very high catalytic activity in the absence of additives. The reaction condition is mild, and the Michael adducts were obtained in very good enantioselectivities (up to 96%), diastereoselectivities (up to 95:5), and chemical yields (up to 95%).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Supelco
Malonic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium malonate dibasic, ≥97.0% (NT)