Merck
Antiproliferative activity of kinetin riboside on HCT-15 colon cancer cell line.
Nucleosides, nucleotides & nucleic acids (2012-06-01)
Mehdi Rajabi, Elena Gorincioi, Enzo Santaniello
ABSTRAKT
Cytokinins and cytokinin nucleosides are purine derivatives with potential anticancer activity both in vitro and in vivo. N(6)-furfuryladenosine (kinetin riboside, KR) displays antiproliferative and apoptogenic activity against various human cancer cell lines and has recently been shown to suppress tumor growth in murine xenograft models of human leukemia and melanoma. In this study, we demonstrate that KR is able to inhibit the proliferation in HCT-15 human colon cancer cells in a dose-dependent manner with a concentration of 2.5 μM, which causes 50% inhibition of cell viability. The cell cycle analysis by flow cytometry showed that KR arrested cell cycle progression in the S Phase by blocking through G(2)/M and G(0)/G(1) phase in HCT-15 colon cells. Moreover, suppression of clonogenic activity occurs after exposure to KR at a concentration of 2.5 μM for HCT-15.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Kinetin, ≥99.0% (HPLC)
Sigma-Aldrich
Kinetin, BioReagent, suitable for plant cell culture, amorphous powder
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline
Sigma-Aldrich
Kinetin solution, 1 mg/mL, BioReagent, suitable for plant cell culture