Merck
  • Fenofibrate increases high-density lipoprotein and sphingosine 1 phosphate concentrations limiting abdominal aortic aneurysm progression in a mouse model.
Fenofibrate increases high-density lipoprotein and sphingosine 1 phosphate concentrations limiting abdominal aortic aneurysm progression in a mouse model.
The American journal of pathology (2012-06-16)
Smriti M Krishna, Sai Wang Seto, Joseph V Moxon, Catherine Rush, Philip J Walker, Paul E Norman, Jonathan Golledge
ABSTRAKT
There are currently no acceptable treatments to limit progression of abdominal aortic aneurysm (AAA). Increased serum concentrations of high-density lipoprotein (HDL) are associated with reduced risk of developing an AAA. The present study aimed to assess the effects of fenofibrate on aortic dilatation in a mouse model of AAA. Male low-density lipoprotein receptor-deficient (Ldlr(-/-)) mice were maintained on a high-fat diet for 3 weeks followed by 6 weeks of oral administration of vehicle or fenofibrate. From 14 to 18 weeks of age, all mice were infused with angiotensin II (AngII). At 18 weeks of age, blood and aortas were collected for assessment of serum lipoproteins, aortic pathology, aortic Akt1 and endothelial nitric oxide synthase (eNOS) activities, immune cell infiltration, eNOS and inducible NOS (iNOS) expression, sphingosine 1 phosphate (S1P) receptor status, and apoptosis. Mice receiving fenofibrate had reduced suprarenal aortic diameter, reduced aortic arch Sudan IV staining, higher serum HDL levels, increased serum S1P concentrations, and increased aortic Akt1 and eNOS activities compared with control mice. Macrophages, T lymphocytes, and apoptotic cells were less evident and eNOS, iNOS, and S1P receptors 1 and 3 were up-regulated in aortas from mice receiving fenofibrate. The present findings suggest that fenofibrate antagonizes AngII-induced AAA and atherosclerosis by up-regulating serum HDL and S1P levels, with associated activation of NO-producing enzymes and reduction of aortic inflammation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sudan IV, Certified Biological Stain Commission,Dye content ≥80 %
Sigma-Aldrich
Sudan IV, certified by the BSC, BioXtra
Supelco
Sudan IV, analytical standard