Merck
Aqueous extract of Artemisia iwayomogi Kitamura attenuates cholestatic liver fibrosis in a rat model of bile duct ligation.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-07-25)
Jong-Min Han, Hyeong-Geug Kim, Min-Kyung Choi, Jin-Suk Lee, Hye-Jung Park, Jing-Hua Wang, Jong-Suk Lee, Seung-Wan Son, Seock-Yeon Hwang, Chang-Gue Son
ABSTRAKT
Cholestatic liver fibrosis, characterized by excessive accumulation of extracellular matrix (ECM) proteins, is associated with bile acid-induced oxidative stress and lipid peroxidation. We evaluated the therapeutic or protective effect of an aqueous extract of Artemisia iwayomogi Kitamura (WAI) in a rat bile duct ligation (BDL)-induced hepatic fibrogenesis model. After BDL, rats were treated once daily with 25 or 50 mg/kg of WAI for 2weeks. The serum bilirubin, aspartate transaminase, alanine transaminase, malondialdehyde, and liver hydroxyproline levels were drastically increased in the BDL group. WAI administration significantly reduced these markers and restored BDL-induced depletion of glutathione content and glutathione peroxidase activity. Cholestatic liver injury and collagen deposition were markedly attenuated by WAI treatment, and these changes were paralleled by significantly suppressed gene and protein expression of fibrogenic factors, including hepatic alphasmooth muscle actin, platelet-derived growth factor, and transforming growth factor β. Our data suggest that WAI may have antifibrotic properties via both improvement of antioxidant activities and inhibition of ECM protein production in the rat model of BDL.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Scopoletin, analytical standard
Sigma-Aldrich
Scopoletin, ≥99%