Merck
  • Synthesis and investigation on photophysical and photochemical properties of 7-oxy-3-methyl-4-phenylcoumarin bearing zinc phthalocyanines.
Synthesis and investigation on photophysical and photochemical properties of 7-oxy-3-methyl-4-phenylcoumarin bearing zinc phthalocyanines.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-07-31)
Mehmet Pişkin, Mahmut Durmuş, Mustafa Bulut
ABSTRAKT
The synthesis of peripherally and non-peripherally tetrakis-(7-oxy-3-methyl-4-phenylcoumarin) and peripherally octakis-[tetrachlorotetra-(7-oxy-3-methyl-4-phenyl coumarin)] substituted zinc(II) phthalocyanines are performed and characterized for the first time in this study. The new compounds show excellent solubility in organic solvents, which makes them candidates for use in different applications. Photophysical (fluorescence quantum yields and lifetimes) and photochemical (singlet oxygen generation and photodegradation under light irradiation) properties of these novel coumarino phthalocyanines are investigated in dimethylformamide (DMF). The effects of the positions (peripheral or nonperipheral) of the substituents on the photophysical and photochemical parameters of the coumarino zinc(II) phthalocyanines are reported. The fluorescence quenching behavior of the studied zinc(II) phthalocyanines by addition of 1,4-benzoquinone (BQ) are also described.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc phthalocyanine, Dye content 97 %