Merck
Interactions of L-Arg with calf thymus DNA using neutral red dye as a fluorescence probe.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-07-31)
Jing Lin, Rutao Liu, Canzhu Gao
ABSTRAKT
The interaction between l-Arg and calf thymus DNA (ctDNA) in sodium acetate-acetic acid buffer (pH=4) was investigated with the use of neutral red (NR) dye as a spectral probe coupled with UV-vis absorption, fluorescence, and circular dichroism (CD) spectroscopy technique. The UV absorption spectroscopy indicated that l-Arg interacted with ctDNA via electrostatic force and the fluorescence enhancing of the DNA-NR system verified the electrostatic interaction. In addition, detectable changes in the CD spectrum of ctDNA in the presence of l-Arg indicated conformational changes in the DNA double helix after interaction with the drug. Docking studies were found to corroborate the experimental results. All these results prove that this drug interacts with ctDNA via an electrostatic binding mode.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Neutral Red, certified by the BSC
Sigma-Aldrich
Neutral Red powder, BioReagent, cell culture | mammalian | 553-24-2, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red, Dye content ≥90 %
Sigma-Aldrich
Neutral Red, suitable for microscopy
Sigma-Aldrich
Neutral Red Solution (0.33%), 3.3 g/L in DPBS, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red Solution, Buffered