Merck
  • High-performance liquid chromatography method for determination of hydrogen peroxide in aqueous solution and application to simulated Martian soil and related materials.
High-performance liquid chromatography method for determination of hydrogen peroxide in aqueous solution and application to simulated Martian soil and related materials.
Environmental monitoring and assessment (2012-08-15)
Spencer M Steinberg
ABSTRAKT
A new method for determination of hydrogen peroxide (H2O2) is described. H2O2 reacts with iodide in the presence of ammonium molybdate and vanillic acid resulting in the formation of iodovanillic acid which is quantified by reversed-phase high-performance liquid chromatography (HPLC) and detected by UV absorption at 280 nm. The method provides a detection limit of ~0.1 μM and is easy to implement. Iodovanillic acid is stable, allowing multiple measurements of the same sample, and the separation power of the HPLC yields high selectivity against potential interferences. This method will be useful for many environmental applications. The method has been applied to quantify the photochemical production of H2O2 by aqueous suspensions of various minerals and soils.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Ammonium molybdate tetrahydrate, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 99.98% trace metals basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland