Merck
  • Cytosolic phospholipase A2 alpha/arachidonic acid signaling mediates depolarization-induced suppression of excitation in the cerebellum.
Cytosolic phospholipase A2 alpha/arachidonic acid signaling mediates depolarization-induced suppression of excitation in the cerebellum.
PloS one (2012-08-29)
De-Juan Wang, Dong Yang, Li-Da Su, Ya-jun Xie, Lin Zhou, Cheng-Long Sun, Yin Wang, Xin-Xin Wang, Liang Zhou, Ying Shen
ABSTRAKT
Depolarization-induced suppression of excitation (DSE) at parallel fiber-Purkinje cell synapse is an endocannabinoid-mediated short-term retrograde plasticity. Intracellular Ca(2+) elevation is critical for the endocannabinoid production and DSE. Nevertheless, how elevated Ca(2+) leads to DSE is unclear. We utilized cytosolic phospholipase A(2) alpha (cPLA(2)α) knock-out mice and whole-cell patch clamp in cerebellar slices to observed the action of cPLA(2)α/arachidonic acid signaling on DSE at parallel fiber-Purkinje cell synapse. Our data showed that DSE was significantly inhibited in cPLA(2)α knock-out mice, which was rescued by arachidonic acid. The degradation enzyme of 2-arachidonoylglycerol (2-AG), monoacylglycerol lipase (MAGL), blocked DSE, while another catabolism enzyme for N-arachidonoylethanolamine (AEA), fatty acid amide hydrolase (FAAH), did not affect DSE. These results suggested that 2-AG is responsible for DSE in Purkinje cells. Co-application of paxilline reversed the blockade of DSE by internal K(+), indicating that large conductance Ca(2+)-activated potassium channel (BK) is sufficient to inhibit cPLA(2)α/arachidonic acid-mediated DSE. In addition, we showed that the release of 2-AG was independent of soluble NSF attachment protein receptor (SNARE), protein kinase C and protein kinase A. Our data first showed that cPLA(2)α/arachidonic acid/2-AG signaling pathway mediates DSE at parallel fiber-Purkinje cell synapse.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Paxilline, powder, ≥98% (HPLC)