Merck
  • Enhanced solubilization and desorption of organochlorine pesticides (OCPs) from soil by oil-swollen micelles formed with a nonionic surfactant.
Enhanced solubilization and desorption of organochlorine pesticides (OCPs) from soil by oil-swollen micelles formed with a nonionic surfactant.
Environmental science & technology (2012-09-25)
Guanyu Zheng, Ammaiyappan Selvam, Jonathan W C Wong
ABSTRAKT
The effect of oil-swollen micelles formed with nonionic surfactant polyoxyethylene sorbitan monooleate (Tween 80), cosurfactant 1-pentanol, and linseed oil on the solubilization and desorption of organochlorine pesticides (OCPs) including DDT and γ-HCH from both loam soil and clay soil were investigated. Results showed that the solubilizing capacities of oil-swollen micelles were dependent on the critical micelle concentration (CMC) of Tween 80. Once the concentrations of oil-swollen micelles exceeded the CMC of Tween 80, the oil-swollen micelles exhibited much higher solubilizing capacity than empty Tween 80 micelles for the two OCPs. Desorption tests revealed that oil-swollen micelles could successfully enhance desorption of OCPs from both loam soil and clay soil. However, compared with the efficiencies achieved by empty Tween 80 micelles, oil-swollen micelles exhibited their superiority to desorb OCPs only in loam soil-water system while was less effective in clay soil-water system. Distribution of Tween 80, 1-pentanol and linseed oil in soil-water system revealed that the difference in the sorption behavior of linseed oil onto the two soils is responsible for the different effects of oil-swollen micelles on the desorption of OCPs in loam soil and clay soil systems. Therefore, oil-swollen micelles formed with nonionic surfactant Tween 80 are better candidates over empty micelle counterparts to desorb OCPs from soil with relatively lower sorption capacity for oil fraction, which may consequently enhance the availability of OCPs in soil environment during remediation processes of contaminated soil.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Pentanol, ACS reagent, ≥99%
Sigma-Aldrich
Linseed Oil
Sigma-Aldrich
1-Pentanol-1-13C, 99 atom % 13C, 99% (CP)
Sigma-Aldrich
1-Pentanol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Supelco
1-Pentanol, analytical standard
Sigma-Aldrich
Amyl alcohol, ≥99%, FG
Sigma-Aldrich
1-Pentanol, ReagentPlus®, ≥99%