Merck
Evaluation of polyurethane based on cellulose derivative-ketoprofen biosystem for implant biomedical devices.
International journal of biological macromolecules (2012-10-06)
Doina Macocinschi, Daniela Filip, Stelian Vlad, Maria Butnaru, Laura Knieling
ABSTRAKT
A polyether-urethane based on polytetrahydrofuran containing hydroxypropyl cellulose for biomedical applications was tested for its biocompatibility. Ketoprofen was incorporated (3% and 6%) in the polyurethane matrix as an anti-inflammatory drug. Dynamic vapour sorption method was employed for testing the water sorption/desorption behaviour of these materials with the determination of the surface isotherms, surface parameters and the kinetic curves of sorption/desorption processes. Cytotoxicity testing in vitro for quantifying cell proliferation was employed, and the results evidence noncytotoxicity for the studied polyurethane-drug systems. In vivo biocompatibility study was performed on 200 g weight male rats. It was found that after implantation of the polyether-urethane samples a reduced acute inflammation occurred, especially for polyurethane samples with added ketoprofen.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoprofen, VETRANAL®, analytical standard
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Ketoprofen, meets USP testing specifications