Merck
Interaction profile of diphenyl diselenide with pharmacologically significant thiols.
Molecules (Basel, Switzerland) (2012-10-23)
Waseem Hassan, Joao Batista Teixeira Rocha
ABSTRAKT
Diphenyl diselenide has shown interesting biological activities in various free-radical-induced damage models and can be considered as a potential candidate drug against oxidative stress. Apart from its anti-oxidant activity, this compound can oxidize various thiols. However there are no detailed studies in the literature about the thiol oxidase-like activity of this compound against biologically significant mono and di-thiols with respect to various pH conditions. Keeping in mind the scarcity of data in this area of organochalcogen chemistry, we report for the first time the kinetics of thiol oxidation by diphenyl diselenide, which was carried out in a commonly used phosphate buffer, not only at physiological pH, but also at a number of acidic values. The relative reactivities of the different thiols with diphenyl diselenide were independent of the pKa of the thiol group, such that at pH 7.4, cysteine and dithiothreitol were the most reactive, while 2,3-dimercapto-1-propanesulfonic acid and glutathione were weakly reactive and extremely low reactivity was observed with dimercaptosuccinic acid. Rate of oxidation was dependent on the pH of the incubation medium. The results obtained will help us in the design of rational strategies for the safe pharmacological use of diphenyl diselenide.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diphenyl diselenide, purum, ≥97.0% (GC)
Sigma-Aldrich
Diphenyl diselenide, 98%