Merck
blaCTX-M-32 on an IncN plasmid in Escherichia coli from beef cattle in the United States.
Antimicrobial agents and chemotherapy (2012-11-21)
Jennifer L Cottell, Neena Kanwar, Lucrecia Castillo-Courtade, Gabhan Chalmers, H Morgan Scott, Bo Norby, Guy H Loneragan, Patrick Boerlin
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Selektywny suplement chlortetracykliny, suitable for microbiology
Sigma-Aldrich
Chlortetracycline hydrochloride, ≥91.0% dry basis (HPLC)
Sigma-Aldrich
Chlortetracycline hydrochloride, BioReagent, suitable for fluorescence, from Streptomyces aureofaciens, ≥85.0% (HPLC)
Supelco
Chlortetracycline hydrochloride, VETRANAL®, analytical standard
Supelco
Ceftiofur hydrochloride, VETRANAL®, analytical standard