Merck
Vibrational spectroscopic and DFT study of trimethoprim.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-12)
Alia Ungurean, Nicolae Leopold, Leontin David, Vasile Chiş
ABSTRAKT
Structural investigations by different vibrational spectroscopic methods: FTIR, FT-Raman and surface-enhanced Raman scattering (SERS) spectroscopy, as well as density functional theory (DFT) calculations were performed on trimethoprim (5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine). A reliable assignment of vibrational IR, Raman and SERS bands was possible by a proper choice of model used in quantum chemical calculations. Based on SERS spectrum analysis it is shown that the molecule is adsorbed on the silver surface through the pyrimidine ring, in a perpendicular orientation. Two theoretical models were used in order to simulate the silver surface and the interaction with trimethoprim molecule, the accuracy of the models being evaluated by comparing the predicted bands position of the two complexes with the SERS result.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trimethoprim, VETRANAL®, analytical standard