Merck
  • Host-guest interaction between new nitrooxoisoaporphine and β-cyclodextrins: synthesis, electrochemical, electron spin resonance and molecular modeling studies.
Host-guest interaction between new nitrooxoisoaporphine and β-cyclodextrins: synthesis, electrochemical, electron spin resonance and molecular modeling studies.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-12)
Fernanda Pérez-Cruz, Benjamín Aguilera-Venegas, Michel Lapier, Eduardo Sobarzo-Sánchez, Eugenio Uriarte Villares, Claudio Olea-Azar
ABSTRAKT
A new nitrooxoisoaporphine derivative was synthetized and characterized by cyclic voltammetry and electron spin resonance. Its aqueous solubility was improved by complexes formation with β-cyclodextrin, heptakis(2,6-di-O-methyl)-β-cyclodextrin and (2-hydroxypropyl)-β-cyclodextrin. In order to assess the inclusion degree reached by nitrooxoisoaporphine in cyclodextris cavity, the stability constants of formation of the complexes were determined by phase-solubility measurements obtaining in all cases a type-A(L) diagram. Moreover, electrochemical studies were carried out, where the observed change in the EPC value indicated a lower feasibility of the nitro group reduction. Additionally, a detailed spatial configuration is proposed for inclusion of derivate within the cyclodextrins cavity by 2D NMR techniques. Finally, these results are further interpreted by means of molecular modeling studies. Thus, theoretical results are in complete agreement with the experimental data.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥98.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥95.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG
Sigma-Aldrich
Heptakis(2,6-di-O-methyl)-β-cyclodextrin