Merck
Relief of hypersensitivity after nerve injury from systemic donepezil involves spinal cholinergic and γ-aminobutyric acid mechanisms.
Anesthesiology (2012-12-12)
Masafumi Kimura, Ken-ichiro Hayashida, James C Eisenach, Shigeru Saito, Hideaki Obata
ABSTRAKT
Evoking spinal release of acetylcholine (ACh) produces antinociception in normal animals and reduces hypersensitivity after nerve injury, and some studies suggest that ACh-mediated analgesia relies on γ-aminobutyric acid (GABA)-ergic signaling in the spinal cord. In this study, the authors tested the spinal mechanisms underlying the antihypersensitivity effects of donepezil, a central nervous system-penetrating cholinesterase inhibitor, in a rat model of neuropathic pain. Male Sprague-Dawley rats were anesthetized, and L5 spinal nerve ligation was performed unilaterally. Withdrawal threshold to a paw pressure test was measured before and after intraperitoneal administration of donepezil, with or without intrathecal antagonists for cholinergic and GABAergic receptors. Microdialysis studies in the ipsilateral dorsal horn of the lumbar spinal cord were also performed to measure extracellular ACh and GABA. Donepezil increased the withdrawal threshold in spinal nerve ligation rats but not in normal rats. The antihypersensitivity effect of donepezil (1 mg/kg) in spinal nerve ligation rats was reduced by intrathecal pretreatment with atropine (30 μg), a muscarinic receptor antagonist; mecamylamine (100 μg), a nicotinic receptor antagonist; bicuculline (0.03 μg), a γ-aminobutyric acid receptor type A antagonist; and CGP 35348 (30 μg), a γ-aminobutyric acid receptor type B antagonist. ACh and GABA concentrations in the microdialysates from the spinal dorsal horn were increased after intraperitoneal donepezil treatment (1 mg/kg) in both normal and spinal nerve ligation rats. Systemic administration of donepezil reduces hypersensitivity after nerve injury by increasing extracellular ACh concentration, which itself induces GABA release in the spinal cord. Activation of this spinal cholinergic-GABAergic interaction represents a promising treatment for neuropathic pain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
CGP 35348 hydrate, ≥97% (NMR), solid