Merck
Quinine-thiourea catalyzed enantioselective hydrophosphonylation of trifluoromethyl 2(1H)-quinazolinones.
Chemical communications (Cambridge, England) (2012-12-19)
Hexin Xie, Aiguo Song, Xinshuai Zhang, Xiaobei Chen, Hao Li, Chunquan Sheng, Wei Wang
ABSTRAKT
An organocatalytic enantioselective addition reaction of cyclic ketoimines with phosphites has been developed for the first time. The process, catalyzed by Soós' quinine thiourea, affords synthetically and medicinally interesting enantioenriched trifluoromethyl dihydroquinazolinones in high yields and with high enantioselectivities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Quinine, 90%
Supelco
Quinine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)