Merck
Conductive polyaniline helixes self-assembled in the absence of chiral dopant.
Chemical communications (Cambridge, England) (2013-01-04)
Chen Li, Jun Yan, Xiujie Hu, Tong Liu, Chenghua Sun, Shizhuo Xiao, Jiantao Yuan, Ping Chen, Shuyun Zhou
ABSTRAKT
The morphological evolution of PANI helixes was achieved without the use of chiral dopants, but by a facile process through the generation of onion-like multi-lamellar vesicles used as a novel soft-template and a liquid crystal acting as a promoter for PANI polymerization and self-assembly, which is a breakthrough for the synthesis of helical structures.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Supelco
Sodium dodecylbenzenesulfonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Dodecylbenzenesulfonic acid solution, 70 wt. % in isopropanol
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium dodecylbenzenesulfonate, technical grade
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium persulfate, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N