Merck
Mechanism of HERG potassium channel inhibition by tetra-n-octylammonium bromide and benzethonium chloride.
Toxicology and applied pharmacology (2013-01-15)
Yan Long, Zuoxian Lin, Menghang Xia, Wei Zheng, Zhiyuan Li
ABSTRAKT
Tetra-n-octylammonium bromide and benzethonium chloride are synthetic quaternary ammonium salts that are widely used in hospitals and industries for the disinfection and surface treatment and as the preservative agent. Recently, the activities of HERG channel inhibition by these compounds have been found to have potential risks to induce the long QT syndrome and cardiac arrhythmia, although the mechanism of action is still elusive. This study was conducted to investigate the mechanism of HERG channel inhibition by these compounds by using whole-cell patch clamp experiments in a CHO cell line stably expressing HERG channels. Tetra-n-octylammonium bromide and benzethonium chloride exhibited concentration-dependent inhibitions of HERG channel currents with IC(50) values of 4nM and 17nM, respectively, which were also voltage-dependent and use-dependent. Both compounds shifted the channel activation I-V curves in a hyperpolarized direction for 10-15mV and accelerated channel activation and inactivation processes by 2-fold. In addition, tetra-n-octylammonium bromide shifted the inactivation I-V curve in a hyperpolarized direction for 24.4mV and slowed the rate of channel deactivation by 2-fold, whereas benzethonium chloride did not. The results indicate that tetra-n-octylammonium bromide and benzethonium chloride are open-channel blockers that inhibit HERG channels in the voltage-dependent, use-dependent and state-dependent manners.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hyamine® 1622 solution, 4 mM in H2O
Sigma-Aldrich
Benzethonium chloride, ≥97% (titration), ≥98% (HPLC)
Sigma-Aldrich
Benzethonium chloride, BioUltra, ≥99.0% (AT)