Merck
Duration of rise in free fatty acids determines salicylate's effect on hepatic insulin sensitivity.
The Journal of endocrinology (2013-01-19)
Sandra Pereira, Wen Qin Yu, María E Frigolet, Jacqueline L Beaudry, Yaniv Shpilberg, Edward Park, Cristina Dirlea, B L Grégoire Nyomba, Michael C Riddell, I George Fantus, Adria Giacca
ABSTRAKT
We have shown in rats that sodium salicylate (SS), which inhibits IkBa kinase B (IKKB), prevents hepatic and peripheral insulin resistance caused by short-term (7  h) i.v. administration of Intralipid and heparin (IH). We wished to further determine whether this beneficial effect of SS persisted after prolonged (48  h) IH infusion, which better mimics the chronic free fatty acid (FFA) elevation of obesity. Hence, we performed hyperinsulinemic euglycemic clamps with tritiated glucose methodology to determine hepatic and peripheral insulin sensitivity in rats infused with saline, IH, IH and SS, or SS alone. SS prevented peripheral insulin resistance (P<0.05) caused by prolonged plasma FFA elevation; however, it did not prevent hepatic insulin resistance. In skeletal muscle, protein levels of phospho-IkBa were augmented by prolonged IH administration and this was prevented by SS, suggesting that IH activates while SS prevents the activation of IKKB. Markers of IKKB activation, namely protein levels of phospho-IkBa and IkBa, indicated that IKKB is not activated in the liver after prolonged FFA elevation. Phosphorylation of serine 307 at insulin receptor substrate (IRS)-1, which is a marker of proximal insulin resistance, was not altered by IH administration in the liver, suggesting that this is not a site of hepatic insulin resistance in the prolonged lipid infusion model. Our results suggest that the role of IKKB in fat-induced insulin resistance is time and tissue dependent and that hepatic insulin resistance induced by prolonged lipid elevation is not due to an IRS-1 serine 307 kinase.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Soybean oil, analytical standard
Sigma-Aldrich
Soybean oil, dietary source of long-chain triglycerides and other lipids
Sigma-Aldrich
Sodium salicylate, 98.0-102.0% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Sodium salicylate, ≥99.5% (HPLC), puriss. p.a.
Sigma-Aldrich
Sodium salicylate, ReagentPlus®, ≥99.5% (titration)
Sigma-Aldrich
Sodium salicylate, puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)