Merck
  • Development, characterization and clinical validation of new sensitive immunofluorometric assay for the measurement of serum thyroglobulin.
Development, characterization and clinical validation of new sensitive immunofluorometric assay for the measurement of serum thyroglobulin.
Arquivos brasileiros de endocrinologia e metabologia (2013-01-19)
Cláudia C D Nakabashi, Rosa Paula M Biscolla, Teresa S Kasamatsu, Teresinha T Tachibana, Rafaela N Barcelos, Eduardo Z Malouf, Danielle M Andreoni, Rui M B Maciel, José Gilberto H Vieira
ABSTRAKT
In the last decade, data published stressed the role of highly-sensitive thyroglobulin (Tg) assays in the follow-up of differentiated thyroid carcinoma (DTC) patients. The present study describes a new, highly-sensitive Tg assay, compares it with an available commercial assay, and validates it in the follow-up of DTC patients. The immunofluorometric high-sensitivity Tg assay is based on monoclonal and polyclonal antibodies produced at our laboratories. It was validated in 100 samples of 87 patients with DTC submitted to total thyroidectomy, 87% of whom also received radioiodine. For correlation, all samples were also tested using a commercial Tg assay (Beckman Access) with functional sensitivity (FS) of 0.1 ng/mL. The new method showed FS of 0.3 ng/mL. The correlation between the two methods was good (r = 0.74; p < 0.0001). The diagnostic sensitivity was 88.9%, and it was increased to 100% when combined with neck US. This new, high-sensitivity Tg assay presented a good correlation with Beckman Access assay and with the clinical outcome of the patients. The continuous availability of a validated assay is an additional advantage for long term follow-up of DTC patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thyroglobulin human
Sigma-Aldrich
Thyroglobulin from porcine thyroid gland, powder