Merck
Antimicrobial resistance in faecal enterococci and Escherichia coli isolates recovered from Iberian wolf.
Letters in applied microbiology (2013-01-23)
A Gonçalves, G Igrejas, H Radhouani, S Correia, R Pacheco, T Santos, R Monteiro, A Guerra, F Petrucci-Fonseca, F Brito, C Torres, P Poeta
ABSTRAKT
The aim of this study was to report the antimicrobial resistance, the molecular mechanisms associated and the detection of virulence determinants within faecal Enterococcus spp. and Escherichia coli isolates of Iberian wolf. Enterococci (n = 227) and E. coli (n = 195) isolates were obtained from faecal samples of Iberian wolf (Canis lupus signatus). High rates of resistance were detected for tetracycline and erythromycin among the enterococci isolates, and most of resistant isolates harboured the tet(M) and/or tet(L) and erm(B) genes, respectively. The blaTEM, tet(A) and/or tet(B), and aadA or strA-strB genes were detected among most ampicillin-, tetracycline- or streptomycin-resistant E. coli isolates, respectively. E. coli isolates were ascribed to phylogroups A (n = 56), B1 (91), B2 (13) and D (35). The occurrence of resistant enterococci and E. coli isolates in the faecal flora of Iberian wolf, including the presence of resistant genes in integrons, and virulence determinants was showed in this study. Iberian wolf might act as reservoir of certain resistance genes that could be spread throughout the environment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Supelco
Streptomycin sesquisulfate hydrate, VETRANAL®, analytical standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water