Merck
  • Thyroglobulin is a sensitive measure of both deficient and excess iodine intakes in children and indicates no adverse effects on thyroid function in the UIC range of 100-299 μg/L: a UNICEF/ICCIDD study group report.
Thyroglobulin is a sensitive measure of both deficient and excess iodine intakes in children and indicates no adverse effects on thyroid function in the UIC range of 100-299 μg/L: a UNICEF/ICCIDD study group report.
The Journal of clinical endocrinology and metabolism (2013-01-25)
Michael B Zimmermann, Isabelle Aeberli, Maria Andersson, Vincent Assey, Jorge A Jara Yorg, Pieter Jooste, Tomislav Jukić, Djoko Kartono, Zvonko Kusić, Eduardo Pretell, Teofilo O L San Luis, Juliawati Untoro, Arnold Timmer
ABSTRAKT
The median urinary iodine concentration (UIC) is a biomarker of iodine intake. According to the World Health Organization, a median UIC in the range 100-199 μg/L indicates adequate and 200-299 μg/L more than adequate intake. Thyroglobulin (Tg) may be a promising functional biomarker of both iodine deficiency and excess. Using a standardized dried blood spots-Tg assay in children, we evaluated the Tg response to both low- and high-iodine intake and estimated the population cutoff point for iodine deficiency or excess. Also, we compared thyroid functions within the UIC ranges of 100-199 vs 200-299 μg/L. We conducted a cross-sectional study in primary schools in 12 countries. SUBJECTS were 6 to 12 years old (n = 2512). We measured UIC, TSH, total T4, Tg, and thyroid antibodies. Over a range of iodine intakes from severely deficient to excessive, Tg concentrations showed a clear U-shaped curve. Compared with iodine-sufficient children, there was a significantly higher prevalence of elevated Tg values in children with iodine deficiency (UIC <100 μg/L) and iodine excess (UIC >300 μg/L). There was no significant change in the prevalence of elevated Tg, TSH, T4, or thyroid antibodies comparing children within the UIC ranges of 100-199 vs 200-299 μg/L. In school-aged children, 1) Tg is a sensitive indicator of both low and excess iodine intake; 2) a median Tg of <13 μg/L and/or <3% of Tg values >40 μg/L indicates iodine sufficiency in the population; 3) the acceptable range of median UIC in monitoring iodized salt programs could be widened to a single category of sufficient iodine intake from 100 to 299 μg/L.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thyroglobulin human
Sigma-Aldrich
Thyroglobulin from porcine thyroid gland, powder