Merck
Allopurinol safely and effectively optimises thiopurine metabolites in patients with autoimmune hepatitis.
Alimentary pharmacology & therapeutics (2013-01-26)
Y S de Boer, N M F van Gerven, N K H de Boer, C J J Mulder, G Bouma, C M J van Nieuwkerk
ABSTRAKT
Ten percent of patients with autoimmune hepatitis (AIH) are nonresponsive or intolerant to thiopurine therapy. A skewed metabolism, leading to the preferential generation of (hepato)toxic thiopurine metabolites (6-MMPs) instead of the metabolic active 6-tioguanine (thioguanine) nucleotides (6-TGNs), may explain this unfavourable outcome. Co-administration of allopurinol to low-dose thiopurine therapy may effectively revert this deviant metabolism, as has been shown in inflammatory bowel disease. To describe the effect of adding allopurinol to low-dose thiopurine therapy in patients with AIH with intolerance or nonresponse to normal thiopurine dosages due to a skewed metabolism. We describe the clinical efficacy and tolerability of allopurinol-thiopurine combination therapy with allopurinol 100 mg and low-dose thiopurine (25-33% of the original dosage) in eight AIH patients with a skewed thiopurine metabolism. Patients were switched because of dose-limiting intolerance (n = 3), nonresponse (n = 3) or loss of response (n = 2) to conventional thiopurine treatment. All eight patients showed biochemical improvement with a reduction in median alanine aminotransferase (ALT) levels of 62 U/L at start to 35 U/L at 1 month (P = 0.03). This clinical benefit was sustained in seven patients. Allopurinol-thiopurine combination therapy effectively bypassed thiopurine side effects in four of five patients. Median 6-tioguanine nucleotides levels increased from 100 to 200 pmol/8 × 10(8) red blood cells (RBC) at 3 months (P = 0.04). Median 6-MMP levels decreased in all patients from 6090 to 175 pmol/8 × 10(8) RBC (P = 0.01). Allopurinol safely and effectively optimises thiopurine therapy in patients with autoimmune hepatitis with intolerance and/or nonresponse due to an unfavourable thiopurine metabolism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Mercaptopurine monohydrate, 98%
Allopurinol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
6-Thioguanine, ≥98%
Sigma-Aldrich
6-Thioguanine, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor