Merck
Investigation of internal structure of fine granules by microtomography using synchrotron X-ray radiation.
International journal of pharmaceutics (2013-02-05)
Shuji Noguchi, Ryusuke Kajihara, Yasunori Iwao, Yukari Fujinami, Yoshio Suzuki, Yasuko Terada, Kentaro Uesugi, Keiko Miura, Shigeru Itai
ABSTRAKT
Computed tomography (CT) using synchrotron X-ray radiation was evaluated as a non-destructive structural analysis method for fine granules. Two kinds of granules have been investigated: a bromhexine hydrochloride (BHX)-layered Celphere CP-102 granule coated with pH-sensitive polymer Kollicoat Smartseal 30-D, and a wax-matrix granule constructed from acetaminophen (APAP), dibasic calcium phosphate dehydrate, and aminoalkyl methacrylate copolymer E (AMCE) manufactured by melt granulation. The diameters of both granules were 200-300 μm. CT analysis of CP-102 granule could visualize the laminar structures of BHX and Kollicoat layers, and also visualize the high talc-content regions in the Kollicoat layer that could not be detected by scanning electron microscopy. Moreover, CT analysis using X-ray energies above the absorption edge of Br specifically enhanced the contrast in the BHX layer. As for granules manufactured by melt granulation, CT analysis revealed that they had a small inner void space due to a uniform distribution of APAP and other excipients. The distribution of AMCE revealed by CT analysis was also found to involve in the differences of drug dissolution from the granules as described previously. These observations demonstrate that CT analysis using synchrotron X-ray radiation is a powerful method for the detailed internal structure analysis of fine granules.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Bromhexine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Bromhexine hydrochloride, ≥98.0% (AT)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~1,000,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~100,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~370,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~80,000, powder, 20 mesh particle size (99% through)
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%