Merck
  • Proteomic biomarkers of preterm birth risk in women with polycystic ovary syndrome (PCOS): a systematic review and biomarker database integration.
Proteomic biomarkers of preterm birth risk in women with polycystic ovary syndrome (PCOS): a systematic review and biomarker database integration.
PloS one (2013-02-06)
Nicolas Galazis, Nikolina Docheva, Kypros H Nicolaides, William Atiomo
ABSTRAKT
Preterm Birth (PTB) is a major cause of neonatal mortality and morbidity. Women with Polycystic Ovary Syndrome (PCOS) are at high risk of PTB. There is a need for research studies to investigate the mechanisms linking PCOS and PTB, to facilitate screening, and develop novel preventative strategies. To list all the proteomic biomarkers of PTB and integrate this list with the PCOS biomarker database to identify commonly expressed biomarkers of the two conditions. A systematic review of PTB biomarkers and update of PCOS biomarker database. All eligible published studies on proteomic biomarkers for PTB and PCOS identified through various databases were evaluated. For the identification of the relevant studies, the following search terms were used: "proteomics", "proteomic", "preterm birth", "preterm labour", "proteomic biomarker" and "polycystic ovary syndrome". This search was restricted to humans only A database on proteomic biomarkers for PTB was created while an already existing PCOS biomarker database was updated. The two databases were integrated and biomarkers that were co-expressed in both women with PCOS and PTB were identified and investigated. A panel of six proteomic biomarkers was similarly differentially expressed in women with PTB and women with PCOS compared to their respective controls (normal age-matched women in the case of PCOS studies and women with term pregnancy in the case of PTB studies). These biomarkers include Pyruvate kinase M1/M2, Vimentin, Fructose bisphosphonate aldolase A, Heat shock protein beta-1, Peroxiredoxin-1 and Transferrin. These proteomic biomarkers (Pyruvate kinase M1/M2, Vimentin, Fructose bisphosphonate aldolase A, Heat shock protein beta-1, Peroxiredoxin-1 and Transferrin) can be potentially used to better understand the pathophysiological mechanisms linking PCOS and PTB. This would help to identify subgroups of women with PCOS at risk of PTB and hence the potential of developing preventative strategies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
Aldolase from rabbit muscle, lyophilized powder, ≥8.0 units/mg protein
Sigma-Aldrich
Aldolase from rabbit muscle, ammonium sulfate suspension, 10-20 units/mg protein