Merck
Attenuated vasodilator effectiveness of protease-activated receptor 2 agonist in heterozygous par2 knockout mice.
PloS one (2013-02-15)
John C Hennessey, John J McGuire
ABSTRAKT
Studies of homozygous PAR2 gene knockout mice have described a mix of phenotypic effects in vitro and in vivo. However, there have been few studies of PAR2 heterozygous (wild-type/knockout; PAR2-HET) mice. The phenotypes of many hemi and heterozygous transgenic mice have been described as intermediates between those of wild-type and knockout animals. In our study we aimed to determine the effects of intermediary par2 gene zygosity on vascular tissue responses to PAR2 activation. Specifically, we compared the vasodilator effectiveness of the PAR2 activating peptide 2-furoyl-LIGRLO-amide in aortas of wild-type PAR2 homozygous (PAR2-WT) and PAR2-HET mice. In myographs under isometric tension conditions, isolated aortic rings were contracted by alpha 1-adrenoeceptor agonist (phenylephrine), and thromboxane receptor agonist (U46619) and then relaxation responses by the additions of 2-furoyl-LIGRLO-amide, acetylcholine, and nitroprusside were recorded. A Schild regression analysis of the inhibition by a PAR2 antagonist (GB-83) of PAR2 agonist-induced aortic ring relaxations was used to compare receptor expression in PAR2-WT to PAR2-HET. PAR2 mRNA in aortas was measured by quantitative real-time PCR. In aortas contracted by either phenylephrine or U46619, the maximum relaxations induced by 2-furoyl-LIGRLO-amide were less in PAR2-HET than in the gender-matched PAR2-WT. GB-83 was 3- to 4-fold more potent for inhibition of 2fly in PAR2-HET than in PAR2-WT. PAR2 mRNA content of aortas from PAR2-HET was not significantly different than in PAR2-WT. Acetylcholine- and nitroprusside-induced relaxations of aortas from PAR2-HET were not significantly different than in PAR2-WT and PAR2 knockout. An interesting secondary finding was that relaxations induced by agonists of PAR2 and muscarinic receptors were larger in females than in males. We conclude that the lower PAR2-mediated responses in PAR2-HET aortas are consistent with evidence of a lower quantity of functional receptor expression, despite the apparently normal PAR2 mRNA content in PAR2-HET aortas.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
R-(-)-Phenylephrine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium nitroprusside dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium nitroferricyanide(III) dihydrate, ACS reagent, ≥99%