Merck
  • Gas chromatography combined with mass spectrometry, flame ionization detection and elemental analyzer/isotope ratio mass spectrometry for characterizing and detecting the authenticity of commercial essential oils of Rosa damascena Mill.
Gas chromatography combined with mass spectrometry, flame ionization detection and elemental analyzer/isotope ratio mass spectrometry for characterizing and detecting the authenticity of commercial essential oils of Rosa damascena Mill.
Rapid communications in mass spectrometry : RCM (2013-02-16)
Federica Pellati, Giulia Orlandini, Katryna A van Leeuwen, Giulia Anesin, Davide Bertelli, Mauro Paolini, Stefania Benvenuti, Federica Camin
ABSTRAKT
The essential oil of Rosa damascena Mill. is known for its fine perfumery application, use in cosmetic preparations and for several pharmacological activities. Due to its high value, it can be easily adulterated with flavors or cheaper oils. This study is aimed at a detailed phytochemical characterization of commercial samples of R. damascena essential oil and at their authenticity assessment. Nineteen commercial samples of R. damascena essential oil of different geographic origin and an additional authentic one, directly extracted by hydro-distillation from fresh flowers, were considered. GC/MS and GC/FID techniques were applied for the phytochemical analysis of the samples. EA/IRMS (Elemental Analyzer/Isotope Ratio Mass Spectrometry) and GC/C (Combustion)/IRMS were used to determine the δ(13)C composition of bulk samples and of some specific components. Citronellol (28.7-55.3%), geraniol (13.5-27.3%) and nonadecane (2.6-18.9%) were the main constituents of Bulgarian and Turkish essential oils, while those from Iran were characterized by a high level of aliphatic hydrocarbons (nonadecane: 3.7-23.2%). The δ(13)C values of bulk samples were between -28.1 and -26.9‰, typical for C3 plants. The δ(13)C values of specific components were in the usual range for natural aromatic substances from C3 plants, except for geranyl acetate, which displayed higher values (up to -18‰). These unusual δ(13)C values were explained by the addition of a natural cheaper oil from a C4 plant (Cymbopogon martinii, palmarosa), which was found to occur in most of the essential oils. GC/C/IRMS, in combination with GC/MS and GC/FID, can be considered as an effective and reliable tool for the authenticity control of R. damascena essential oil.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Supelco
Geraniol, analytical standard
Sigma-Aldrich
(S)-(−)-β-Citronellol, ≥97%
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
(R)-(+)-β-Citronellol, 95%
Supelco
(−)-β-Citronellol, analytical standard
Supelco
(±)-β-Citronellol, analytical standard
Sigma-Aldrich
β-Citronellol, 95%
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
Sigma-Aldrich
Nonadecane, 99%
Sigma-Aldrich
Citronellol, ≥95%, FCC, FG
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
Supelco
Nonadecane, analytical standard
Sigma-Aldrich
Citronellol, mixture of isomers, natural, ≥95%, FG