Merck
Total phenolic content and antioxidative properties of commercial tinctures obtained from some Lamiaceae plants.
Natural product communications (2013-02-19)
Adam Kowalczyk, Izabela Biskup, Izabela Fecka
ABSTRAKT
The antioxidant level of commercial tinctures from three Lamiaceae plants, Salvia officinalis, Mentha piperita, and Melissa officinalis, have been determined by the Folin-Ciocalteu method, the 2.2-diphenyl-2-picrylhydracyl hydrate (DPPH) radical scavenging technique and ABTS assay. Total phenolic content was expressed as GAE (gallic acid equivalent) and ranged from 0.24 to 3.99 mg/mL. Antioxidant activity in the ABTS assay, calculated as TEAC (Trolox equivalent antioxidant capacity), ranged from 23.5 to 35.6 micromol Trolox/mL, while in the DPPH method, the EC50 value ranged from 0.04 to 0.07 mL/assay. Radical scavenging activity was correlated with total phenolic content. Correlations between ABTS and F-C methods, DPPH and F-C methods and ABTS and DPPH methods were calculated. The obtained results can be useful as additional information about the antioxidant activity of galenical preparations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)