Merck
Comparative study on the responses of concentrations of B₆-vitamers in several tissues of mice to the dietary level of pyridoxine.
Journal of nutritional science and vitaminology (2013-02-20)
Kabo Masisi, Sofya Suidasari, Peipei Zhang, Yukako Okazaki, Noriyuki Yanaka, Norihisa Kato
ABSTRAKT
To examine the responses of the levels of B₆-vitamers in several tissues to the dietary level of pyridoxine (PN), mice were fed diets containing 0, 1, 7 (the recommended level) or 35 mg PN HCl/kg diet for 5 wk. Compared with the 0 mg PN HCl/kg diet, the 35 mg PN HCl/kg diet caused the highest elevation in the concentration of pyridoxal 5'-phosphate (PLP) in small intestine and epididymal adipose tissue, moderate elevation in colon, lung, spleen and stomach, slight elevation in brain, kidney and liver (p<0.05), and no elevation in heart and gastrocnemius muscle. In general, the alterations in PLP level in many tissues and serum were remarkable for diets between 1 mg and 7 mg PN HCl/kg diets. Compared to the 7 mg PN HCl/kg diet, the 35 mg PN HCl/kg diet further elevated the PLP level in adipose tissue, spleen and stomach (p<0.05). Dietary supplemental PN elevated the level of PN in small intestine and colon in a dose-dependent manner (p<0.05), but not in other tissues. There was a significant correlation between the PN and PLP levels in small intestine and colon (p<0.05), implying that PN absorbed from the diet can be at least in part metabolized to PLP within the absorptive intestinal cells. The results suggest that the responses of concentrations of B₆-vitamers to dietary level of PN greatly differ among several tissues.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyridoxine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Pyridoxine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridoxine, ≥98%