Merck
Acute disseminated encephalomyelitis associated with acute Toxoplasma gondii Infection.
Pediatric neurology (2013-02-20)
Ayse Aksoy, Gonul Tanir, Mehpare Ozkan, Melek Oguz, Yasemin Tasci Yıldız
ABSTRAKT
Acute disseminated encephalomyelitis is an acute demyelinating disorder of the central nervous system, which principally affects the brain and spinal cord. It usually follows a benign infection or vaccination in children. Although a number of infectious agents have been implicated in acute disseminated encephalomyelitis, Toxoplasma gondii infection has not been described previously in children. Acquired T. gondii infection presents with lymphadenopathy and fever and usually spontaneously resolves in immunocompetent patients. We describe a previously healthy 10-year-old boy with acute disseminated encephalomyelitis associated with acute acquired Toxoplasma gondii infection, the symptoms of which initially began with nuchal stiffness, difficulty in walking, and urinary and stool incontinence; he later had development of motor and sensory impairment in both lower extremities and classical magnetic resonance imaging lesions suggestive of the disease. The patient recovered completely after the specific therapy for acquired T. gondii infection and pulse prednisolone. Although acute acquired Toxoplasma gondii infection has not been reported previously in association with acute disseminated encephalomyelitis, clinicians should keep in mind this uncommon cause of a common disease when evaluating a patient with acute disseminated encephalomyelitis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Supelco
Pyrimethamine, VETRANAL®, analytical standard
Pyrimethamine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%