Merck
Biomechanical properties of ciprofloxacin loaded bone cement.
Acta orthopaedica et traumatologica turcica (2013-04-04)
Talip Teoman Aslan, Zekeriya Öztemür, Mahmut Çifçi, Gündüz Tezeren, Hayati Öztürk, Okay Bulut
ABSTRAKT
The purpose of this study was to investigate the biomechanical properties of bone cement used in joint replacement surgery after the addition of ciprofloxacin. The first group received bone cement only and served as a control for the 4 groups where 500 mg, 1000 mg, 1500 mg and 2000 mg of ciprofloxacin were added to yield 40 g of bone cement. Axial compression tests were conducted using a 50,000 Newton capacity tension-compression testing device. While axial compression strength at failure was 80.2±4.3 MPa in the control group, values in the ciprofloxacin-treated groups decreased with rising concentration of ciprofloxacin to 74.5±5.4 MPa, 70.6±4.8 MPa, 70.5±4.7 MPa, and 69.3±3.4 MPa. Bone cement with addition of 500 to 1500 mg ciprofloxacin maintained mechanical axial strength values above 70.0 MPa recommended by American Society for Testing and Materials and can be safely used in joint replacement surgery.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard