Merck
Low glutamine levels during critical illness--adaptive or maladaptive?
The New England journal of medicine (2013-04-19)
Greet Van den Berghe
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland